Learn More
Cml Supply CML Supply 20ga x 0.5" Yellow Blunt Tip Dispensing Fill Needle 500/pack
Catalog No. NC1582199
Encompass_Preferred
20 Gauge 1/2" length Blunt Tip Stainless Steel Dispense Needles. Fits all standard Luer Lock or Luer Slip Syringes. Compatible with EFD, Nordson, Techon, PVA dispensing Systems. UV safe plastic hub with luer taper. Non-sterile, but may be sterilized by autoclave. For Industrial use only.
Catalog No. NC1582199 Supplier Cml Supply Supplier No. 90120050D
20 Gauge 1/2" length Blunt Tip Stainless Steel Dispense Needles. Fits all standard Luer Lock or Luer Slip Syringes. Compatible with EFD, Nordson, Techon, PVA dispensing Systems. UV safe plastic hub with luer taper. Non-sterile, but may be sterilized by autoclave. For Industrial use only.