Learn More
Grainger Closed Head Transport Drums, 20 gal
Catalog No. NC1965691
Encompass_Preferred
The 20 gallon closed head transport drum is made of high density polyethylene with a 20 gallon capacity. It features a 2 inch NPS threaded bung closure, is UN rated for hazardous materials, has four rolling hoops for mobility, and dimensions of 23.5 inches diameter x 34.5 inches high. The drum has a natural white color and offers corrosion and chemical resistance. It is suitable for shipping and storing liquid materials. The estimated weight is 22 pounds when empty.
Catalog No. NC1965691 Supplier Grainger Supplier No. 23Y343
The 20 gallon closed head transport drum is made of high density polyethylene with a 20 gallon capacity. It features a 2 inch NPS threaded bung closure, is UN rated for hazardous materials, has four rolling hoops for mobility, and dimensions of 23.5 inches diameter x 34.5 inches high. The drum has a natural white color and offers corrosion and chemical resistance. It is suitable for shipping and storing liquid materials. The estimated weight is 22 pounds when empty.