Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 20-Hc-me-pyrrolidine | 00-00-0 | 99.7% | 429.68 g/mol | C28H47NO2 | 5 MG
SDP

Catalog No. 5000221245
Encompass_Preferred

20-HC-Me-Pyrrolidine is a small-molecule inhibitor of Aster proteins that blocks Aster-mediated cholesterol binding and transfer and inhibits LDL-derived cholesterol movement to the endoplasmic reticulum. Supplied as a high-purity research chemical for biochemical and cellular studies.

  • Potent inhibitor of Aster proteins with reported IC50s Aster-A 0.11 μM, Aster-B 0.06 μM, Aster-C 0.71 μM.
  • High purity suitable for research applications (reported ~99.69%).
  • Well-characterized solubility in DMSO (≈6.67 mg/mL) for stock preparations.
  • Recommended storage at 4°C; stock solutions stable at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C28H47NO2; molecular weight 429.68 g/mol.

Catalog No. 50-002-21245 Supplier Medchemexpress LLC Supplier No. HY1450015MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

20-HC-Me-Pyrrolidine is a small-molecule inhibitor of Aster proteins that blocks Aster-mediated cholesterol binding and transfer and inhibits LDL-derived cholesterol movement to the endoplasmic reticulum. Supplied as a high-purity research chemical for biochemical and cellular studies.

  • Potent inhibitor of Aster proteins with reported IC50s Aster-A 0.11 μM, Aster-B 0.06 μM, Aster-C 0.71 μM.
  • High purity suitable for research applications (reported ~99.69%).
  • Well-characterized solubility in DMSO (≈6.67 mg/mL) for stock preparations.
  • Recommended storage at 4°C; stock solutions stable at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C28H47NO2; molecular weight 429.68 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.