Learn More
Medchemexpress LLC 20-hc-me-pyrrolidine | 00-00-0 | 99.7% | C28H47NO2 | 10MG
SDP
Catalog No. 5000261902
Encompass_Preferred
20-HC-Me-Pyrrolidine is a small-molecule research reagent that inhibits Aster proteins (Aster-A, Aster-B, Aster-C). It is provided as a solid with high reported purity and characterized potency for in vitro biochemical studies; solvent and storage guidelines support reliable handling in research workflows.
  • Potent inhibitor of Aster proteins: IC50 0.11 μM (Aster-A), 0.06 μM (Aster-B), 0.71 μM (Aster-C).
  • High purity (99.69%) suitable for biochemical assays.
  • Soluble in DMSO at 6.67 mg/mL (15.52 mM) for stock solutions.
  • Stable when protected from light and stored under inert atmosphere per handling instructions.
  • Available in small-scale quantities to support discovery and assay development.
Catalog No. 50-002-61902 Supplier Medchemexpress LLC Supplier No. HY14500110MG
Add to Cart
Add to Cart
20-HC-Me-Pyrrolidine is a small-molecule research reagent that inhibits Aster proteins (Aster-A, Aster-B, Aster-C). It is provided as a solid with high reported purity and characterized potency for in vitro biochemical studies; solvent and storage guidelines support reliable handling in research workflows.
  • Potent inhibitor of Aster proteins: IC50 0.11 μM (Aster-A), 0.06 μM (Aster-B), 0.71 μM (Aster-C).
  • High purity (99.69%) suitable for biochemical assays.
  • Soluble in DMSO at 6.67 mg/mL (15.52 mM) for stock solutions.
  • Stable when protected from light and stored under inert atmosphere per handling instructions.
  • Available in small-scale quantities to support discovery and assay development.