Learn More
Cayman Chemical 20-HETE, 79551-86-3, 100 µg
SDP
Supplier:  Cayman Chemical 90030100
Encompass_Preferred
A CYP450 metabolite of arachidonic acid in the renal and cerebral vasculature; exhibits vasoconstrictor activity
Catalog No. NC9935387