Learn More
Cayman Chemical 20-hydroxy Ecdysone, Purity: ≥98%, CAS: 5289-74-7, 50 mg
SDP
Catalog No. 502970338
Encompass_Preferred
An ecdysteroid hormone produced in arthropod species that induces developmental changes associated with ecdysis and the completion of metamorphosis; used as a tool in inducible gene regulation systems controlled by reporter cells transfected with the ecdysone receptor
Catalog No. 50-297-0338 Supplier Cayman Chemical Supplier No. 1614550 MG
An ecdysteroid hormone produced in arthropod species that induces developmental changes associated with ecdysis and the completion of metamorphosis; used as a tool in inducible gene regulation systems controlled by reporter cells transfected with the ecdysone receptor