Learn More
Cayman Chemical 20-hydroxy LeukotrIen B4 100ug
SDP
Catalog No. 502984701
Encompass_Preferred
A metabolite of LTB4 in human neutrophils; inhibits LTB4-induced degranulation of human neutrophilis (Ki = 13.3 nM)
Catalog No. 50-298-4701 Supplier Cayman Chemical Supplier No. 20190100 UG
A metabolite of LTB4 in human neutrophils; inhibits LTB4-induced degranulation of human neutrophilis (Ki = 13.3 nM)