Learn More
Abcam 20-Hydro x yecdysone (Ecdysterone), Steroid invertebrate hormone, 10MG
Catalog No. 502706245
Encompass_Preferred
MW 480.6 Da, Purity >98%. Steroid invertebrate hormone. Regulates molting, metamorphosis and reproduction in arthropods. Neuroprotective against oxidative stress in mammals. Inhibits NF-κB activation and iNOS expression. Antioxidant and apoptotic agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6245 Supplier Abcam Supplier No. AB14242510MG
MW 480.6 Da, Purity >98%. Steroid invertebrate hormone. Regulates molting, metamorphosis and reproduction in arthropods. Neuroprotective against oxidative stress in mammals. Inhibits NF-κB activation and iNOS expression. Antioxidant and apoptotic agent.
The product is subject to the following: Abcam Restricted Use Statement