Learn More
Arbor Assays 20-Hydroxyecdysone ELISA Kit (5 Plate)
SDP
Catalog No. 502278500
Encompass_Preferred
The DetectX 20-Hydroxyecdysone ELISA Kits measure 20-hydroxyecdysone in arthropods and plants.
This item is not returnable. View return policy
Catalog No. 50-227-8500 Supplier Arbor Assays Supplier No. K066H5
The DetectX 20-Hydroxyecdysone ELISA Kits measure 20-hydroxyecdysone in arthropods and plants.