Learn More
Arbor Assays 20-Hydroxyecdysone ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K066H1
Encompass_Preferred
The DetectX 20-Hydroxyecdysone ELISA Kits measure 20-hydroxyecdysone in arthropods and plants.
Catalog No. 50-227-8499