Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 20-Hydroxyecdysone ELISA Kit

Catalog No. EIAHYDX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAHYDX10 10 x 96 Tests
EIAHYD 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIAHYDX10 Supplier Invitrogen™ Supplier No. EIAHYDX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

The 20-Hydroxyecdysone Competitive ELISA quantitates 20-Hydroxyecdysone (20E) in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The 20-Hydroxyecdysone Competitive ELISA research-use-only kit is designed to quantitatively measure 20-Hydroxyecdysone independent of species. A 20-Hydroxyecdysone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit 20-Hydroxyecdysone antibodies and 20-Hydroxyecdysone-peroxidase conjugate is added to the wells. The 20-Hydroxyecdysone-peroxidase conjugate and any 20-Hydroxyecdysone in the sample will compete to bind to the anti-rabbit 20-Hydroxyecdysone antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound 20-Hydroxyecdysone-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of 20-Hydroxyecdysone present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

20-Hydroxyecdysone is the most widely occurring ecdysteroid in both plant and animal species. It controls cell death during metamorphosis of Drosophila melanogaster.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 20 E; C27H44O7; ecdysterone
Assay Range 102.4-25000 pg/mL
Assay Sensitivity 197.8 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 10 x 96 Assays
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 20E; C27H44O7; ecdysterone20E; C27H44O7; ecdysterone;
Interassay CV 9%
Intraassay CV 10%
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target 20-Hydroxyecdysone
Test Time 45 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.