Learn More
Medchemexpress LLC 20-hydroxyeicosanoic acid | 62643-46-3 | MFCD02259050 | >98.0% | 328.53 g/mol | C20H40O3 | 250MG
SDP
Catalog No. 5000238557
Encompass_Preferred
20-Hydroxyeicosanoic acid is an omega-hydroxylated C20 saturated fatty acid used as a biochemical standard and research reagent in lipidomics, analytical chemistry, and synthesis. It has molecular formula C20H40O3, molecular weight 328.53 g/mol, CAS 62643-46-3, and is typically supplied as a high-purity solid for analytical use.
  • Omega-hydroxylated C20 fatty acid for lipid research.
  • High purity suitable for analytical and biochemical assays.
  • Available in small milligram to gram quantities for method development.
  • Suitable as a reference standard or synthesis intermediate.
  • Solid form provides convenient storage and handling.
Catalog No. 50-002-38557 Supplier Medchemexpress LLC Supplier No. HY124311250MG
20-Hydroxyeicosanoic acid is an omega-hydroxylated C20 saturated fatty acid used as a biochemical standard and research reagent in lipidomics, analytical chemistry, and synthesis. It has molecular formula C20H40O3, molecular weight 328.53 g/mol, CAS 62643-46-3, and is typically supplied as a high-purity solid for analytical use.
  • Omega-hydroxylated C20 fatty acid for lipid research.
  • High purity suitable for analytical and biochemical assays.
  • Available in small milligram to gram quantities for method development.
  • Suitable as a reference standard or synthesis intermediate.
  • Solid form provides convenient storage and handling.