Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 20-hydroxyeicosanoic acid | 62643-46-3 | MFCD02259050 | 98.0% | 328.54 g/mol | C20H40O3 | 10G
SDP

Catalog No. 5000238554
Encompass_Preferred

20-Hydroxyeicosanoic acid is a hydroxylated long-chain (C20) fatty acid supplied as a high-purity research reagent for biochemical and analytical applications. It is suitable as a reference standard for metabolic studies and for analytical method development.

  • High purity 98.0%
  • Molecular formula C20H40O3; molecular weight 328.54 g/mol
  • Available in 10 g packaging
  • Storage: room temperature (3 years); in solvent: -80°C (2 years), -20°C (1 year)
  • Intended for research and analytical use; not for human or clinical use

Catalog No. 50-002-38554 Supplier Medchemexpress LLC Supplier No. HY12431110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

20-Hydroxyeicosanoic acid is a hydroxylated long-chain (C20) fatty acid supplied as a high-purity research reagent for biochemical and analytical applications. It is suitable as a reference standard for metabolic studies and for analytical method development.

  • High purity 98.0%
  • Molecular formula C20H40O3; molecular weight 328.54 g/mol
  • Available in 10 g packaging
  • Storage: room temperature (3 years); in solvent: -80°C (2 years), -20°C (1 year)
  • Intended for research and analytical use; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.