Learn More
Analytical Sales & Services 20µL SS Sample Loop for 7125 & 7010 (471254 & 470104) SS Manual Injector Valves
Catalog No. 5000804957
Encompass
20µL Stainless Steel Sample Loop for 7125 & 7010 (471254 & 470104) Stainless Steel Manual Injector Valves
Qty/Pkg: EACH
Catalog No. 50-008-04957 Supplier Analytical Sales & Services Supplier No. 470224
20µL Stainless Steel Sample Loop for 7125 & 7010 (471254 & 470104) Stainless Steel Manual Injector Valves
Qty/Pkg: EACH