Learn More
California Pacific Lab & Consulting 20 Liter Rectangular Carboy, HDPE, 70mm Cap Size (Nalgene)
SDP
Catalog No. NC1802734
Encompass_Preferred
Space saving rectangular carboy constructed of HDPE with polypropylene screw closure. Sturdy handle attached to molded in shoulder loops. Molded-in graduations of 1 gallon and 5Ltr. Increments. Wide mouth for easy filling. For use with EF-4-30020 and EF-30020. 399mm H x 218mm L x 318mm W (15.7" H x 8.6" L x 12.5" W)
Catalog No. NC1802734 Supplier California Pacific Lab & Consulting Supplier No. EC20020
Space saving rectangular carboy constructed of HDPE with polypropylene screw closure. Sturdy handle attached to molded in shoulder loops. Molded-in graduations of 1 gallon and 5Ltr. Increments. Wide mouth for easy filling. For use with EF-4-30020 and EF-30020. 399mm H x 218mm L x 318mm W (15.7" H x 8.6" L x 12.5" W)