Learn More
Quality Lab Accessories 20 Mesh Clip Style Basket, 316 Stainless Steel, Electropolished, 40 pcs
Catalog No. NC3451213
Encompass
The 20 Mesh Clip Style Basket from Quality Lab Accessories is engineered for precision and reliability in dissolution testing. Its 20 mesh screen ensures consistent sample containment and accurate dissolution profiles, providing high-quality performance for scientific applications. The clip-style design simplifies attachment within dissolution apparatus, setting it apart from other baskets. This durable accessory is made from 316 stainless steel and meets ISO9001 standards, ensuring it upholds exceptional quality and compliance.
Catalog No. NC3451213 Supplier Quality Lab Accessories Supplier No. BSK02001
The 20 Mesh Clip Style Basket from Quality Lab Accessories is engineered for precision and reliability in dissolution testing. Its 20 mesh screen ensures consistent sample containment and accurate dissolution profiles, providing high-quality performance for scientific applications. The clip-style design simplifies attachment within dissolution apparatus, setting it apart from other baskets. This durable accessory is made from 316 stainless steel and meets ISO9001 standards, ensuring it upholds exceptional quality and compliance.