Learn More
Dual Manufacturing Company Inc Market Grade Sieve, #20 Mesh
Catalog No. NC0957171
Encompass_Preferred
Mesh Openings: .034 in, .86 mm. Diameter: 5 in.
Catalog No. NC0957171 Supplier Dual Manufacturing Company Inc Supplier No. MG520
Mesh Openings: .034 in, .86 mm. Diameter: 5 in.