Learn More
Antron Engineering 20 ML H.P.SYRINGE W/LUER-LOK
SDP
Catalog No. 50871832
Encompass_Preferred
Standard Syringe with Luer-Lock Fitting. These syringes are intended for high-pressure applications and offer good resistance to most aggressive liquids. Syringe barrels and plungers are made of type 316 stainless steel. Syringes are leak-free to pressures up to 750psi, except for 8mL syringes that are leak-free to pressures up to 1500psi. They fit most syringe pumps. Autoclavable.
Catalog No. 50-871-832 Supplier Antron Engineering Supplier No. PRO7300LL
Standard Syringe with Luer-Lock Fitting. These syringes are intended for high-pressure applications and offer good resistance to most aggressive liquids. Syringe barrels and plungers are made of type 316 stainless steel. Syringes are leak-free to pressures up to 750psi, except for 8mL syringes that are leak-free to pressures up to 1500psi. They fit most syringe pumps. Autoclavable.