Learn More
HEPATOCHEM 20 ml quartz vial
Catalog No. NC3781897
Encompass_Preferred
20 ml quartz vials 28x61mm with 24-400 thread (cap with septum included)
Catalog No. NC3781897 Supplier HEPATOCHEM Supplier No. HCK102101054
20 ml quartz vials 28x61mm with 24-400 thread (cap with septum included)