Learn More
Agilent Technologies 20 ML VIAL KIT W CAP & SEPTA
Catalog No. NC1785710
Encompass
Vial kit, crimp top, headspace, clear, 20 mL, silver aluminum caps with safety feature, certified, flat bottom, molded PTFE/black butyl septa, 100/pk. Vial size: 22.75 x 75 mm (20 mm cap)
This item is not returnable. View return policy
Catalog No. NC1785710 Supplier Agilent Technologies Supplier No. 51820839
Vial kit, crimp top, headspace, clear, 20 mL, silver aluminum caps with safety feature, certified, flat bottom, molded PTFE/black butyl septa, 100/pk. Vial size: 22.75 x 75 mm (20 mm cap)