Learn More
Medchemexpress LLC 20 mM HEPES, 150 mM NaCl, 240 mM Sucrose, pH 7.2 buffer | 10 ML
SDP
Catalog No. 5000375376
Encompass_Preferred
This physiological buffer is composed of 20 mM HEPES, 150 mM NaCl, and 240 mM Sucrose, meticulously adjusted to a pH of 7.2. It is specifically designed for the resuspension of human erythrocyte ghost preparations.
  • Physiological buffer
  • Composed of 20 mM HEPES, 150 mM NaCl, and 240 mM Sucrose
  • Adjusted to pH 7.2
  • Used for resuspending human erythrocyte ghost preparations
Catalog No. 50-003-75376 Supplier Medchemexpress LLC Supplier No. HY15035010ML
This physiological buffer is composed of 20 mM HEPES, 150 mM NaCl, and 240 mM Sucrose, meticulously adjusted to a pH of 7.2. It is specifically designed for the resuspension of human erythrocyte ghost preparations.
  • Physiological buffer
  • Composed of 20 mM HEPES, 150 mM NaCl, and 240 mM Sucrose
  • Adjusted to pH 7.2
  • Used for resuspending human erythrocyte ghost preparations