Learn More
Starna Cells Inc 20mm Colorimeter cells 55 x 24 x 56 mm
SDP
Catalog No. NC9626507
Encompass_Preferred
Colorimeter cells for Hunter colorimeter, Glass cell with wide and tall window, Use:Transmission/Absorbtion measurement, Clear windows:2, Open top, Path Length :20mm, Exterior (W x L x H): 55 x 24 x 56 mm, Interior (W x L): 51 x 20 mm
Catalog No. NC9626507 Supplier Starna Cells Inc Supplier No. 93G20
Colorimeter cells for Hunter colorimeter, Glass cell with wide and tall window, Use:Transmission/Absorbtion measurement, Clear windows:2, Open top, Path Length :20mm, Exterior (W x L x H): 55 x 24 x 56 mm, Interior (W x L): 51 x 20 mm