Learn More
Advalue Technologies Alumina Crucible High Form
SDP
Catalog No. NC1882648
Encompass
AL-1750 Alumina Crucible High Form - Capacity 750ml - Top Dia 112mm - Bottom Dia 70mm - Height 132mm
Catalog No. NC1882648 Supplier Advalue Technologies Supplier No. AL1750
AL-1750 Alumina Crucible High Form - Capacity 750ml - Top Dia 112mm - Bottom Dia 70mm - Height 132mm