Learn More
Electron Microscopy Sciences 20% Paraformaldehyde (formaldehyde) aqueous solution, 10x10 mL
SDP
Catalog No. 50980492
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 10 mL ampoules sealed under inert gas.
This item is not returnable. View return policy
Catalog No. 50-980-492 Supplier Electron Microscopy Sciences Supplier No. 15713
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 10 mL ampoules sealed under inert gas.