Learn More
Electron Microscopy Sciences 20% Paraformaldehyde (formaldehyde) aqueous solution, 100 mL
SDP
Catalog No. 50980493
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.
This item is not returnable. View return policy
Catalog No. 50-980-493 Supplier Electron Microscopy Sciences Supplier No. 15713S
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.