Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Pesticide Mix Standard, 20 Component, 10 μg/mL, 1 mL

Catalog No. 50466288 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This 20 Pesticide Mix Standard simplifies pesticide analysis by providing a pre-mixed solution, reducing the need for individual measurement. It assists researchers in accelerating the verification and validation of pesticide screening techniques. The standard ensures accurate formulation and unwavering traceability, which is crucial for data comparability and consistent preparation of reference materials.

  • Contains 20 pesticides at 10 μg/mL each in Acetonitrile for simplified analysis.
  • Formulated as a liquid reference standard.
  • Suitable for LCMS detection and Reversed Phase separation mode.
  • Store at 4°C (Fridge) with a shelf life of 36 Months.
  • Shipped under ambient conditions.
  • Volume: 1 mL.
  • Application for pesticide analysis.
  • One unit per package.
  • Hazardous material requiring special freight handling.

Catalog No. 50-466-288 Supplier Waters Corp Supplier No. 186006348
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 20 Pesticide Mix Standard simplifies pesticide analysis by providing a pre-mixed solution, reducing the need for individual measurement. It assists researchers in accelerating the verification and validation of pesticide screening techniques. The standard ensures accurate formulation and unwavering traceability, which is crucial for data comparability and consistent preparation of reference materials.

  • Contains 20 pesticides at 10 μg/mL each in Acetonitrile for simplified analysis.
  • Formulated as a liquid reference standard.
  • Suitable for LCMS detection and Reversed Phase separation mode.
  • Store at 4°C (Fridge) with a shelf life of 36 Months.
  • Shipped under ambient conditions.
  • Volume: 1 mL.
  • Application for pesticide analysis.
  • One unit per package.
  • Hazardous material requiring special freight handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.