Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific 20-Place Slide Tray Cardboard with Trim

Catalog No. 14127428
Change view
Click to view available options
Color:
Black
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-428 Black
14-127-367 Red
14-127-418 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-127-428 Supplier Heathrow Scientific Supplier No. HS9902
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cardboard slide trays hold standard slides (76 x 26 mm and 3 x 1 in.)

  • Organize and Protect
    • Index on the front cover corresponds with the number grid inside for identification
    • Recessed slide compartments and inside covers for specimen protection
    • Slides cannot be accessed when cover is closed reducing contamination
  • Sturdy Construction
    • Compartment holder is made from a double layer of densely pressed cardboard
    • Cover is single layer cardboard
    • Front protection flaps are attached with durable tape

Specifications

Type Slide Storage Tray
Slide Capacity 20
Color Black
Dimensions (L x W x H) 8.2 x 13.4 x 0.4 in. (20.8 x 34.1 x 0.9 cm)
Material Cardboard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.