Learn More
Zymo Research Corporation EZ-Vac Vacuum Manifold
SDP
Catalog No. NC0994627
Encompass_Preferred
The vacuum manifold allows researchers to simplify their nucleic acid purification workflows further by eliminating the need for multiple centrifugation steps and disposal of flow-through from collection tubes.
Catalog No. NC0994627 Supplier Zymo Research Corporation Supplier No. S7000
The vacuum manifold allows researchers to simplify their nucleic acid purification workflows further by eliminating the need for multiple centrifugation steps and disposal of flow-through from collection tubes.