Learn More
Medchemexpress LLC HY-N1401 5mg Medchemexpress, 20(R)-Ginsenoside Rh2 CAS:112246-15-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14015MG
Encompass_Preferred
Medchemexpress, HY-N1401 5mg 20(R)-Ginsenoside Rh2 CAS:112246-15-8 20(R)-Ginsenoside Rh2, a matrix metalloproteinase (MMP) inhibitor, acts as a cell antiproliferator. It has anticancer effects via blocking cell proliferation and causing G1 phase arrest. 20(R)-Ginsenoside Rh2 induces apoptosis, and has anti-inflammatory and antioxidative activity[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0334