Learn More
Cayman Chemical 20 S-GInsenosIde Rg3 10mg
SDP
Catalog No. 502973141
Encompass_Preferred
A panaxadiol that has diverse in vitro and in vivo effects, including anti-cancer, neuroprotective, anti-hypertensive, and anti-inflammatory actions; induces apoptosis and inhibits angiogenesis in a variety of cancer models
Catalog No. 50-297-3141 Supplier Cayman Chemical Supplier No. 1533210 MG
A panaxadiol that has diverse in vitro and in vivo effects, including anti-cancer, neuroprotective, anti-hypertensive, and anti-inflammatory actions; induces apoptosis and inhibits angiogenesis in a variety of cancer models