Learn More
Cayman Chemical 20(S)-Ginsenoside Rg3, Purity: ≥95%, CAS: 14197-60-5, 50 mg
SDP
Catalog No. 502968787
Encompass_Preferred
A panaxadiol that has diverse in vitro and in vivo effects, including anti-cancer, neuroprotective, anti-hypertensive, and anti-inflammatory actions; induces apoptosis and inhibits angiogenesis in a variety of cancer models
Catalog No. 50-296-8787 Supplier Cayman Chemical Supplier No. 1533250 MG
A panaxadiol that has diverse in vitro and in vivo effects, including anti-cancer, neuroprotective, anti-hypertensive, and anti-inflammatory actions; induces apoptosis and inhibits angiogenesis in a variety of cancer models