Learn More
Cayman Chemical 20 S-GInsenosIde Rh2 10mg
SDP
Catalog No. 502987855
Encompass_Preferred
A natural steroid glycoside with diverse effects; inhibits release of β-hexosaminidase from RBL-2H3 cells (IC50 = 100 μM) and inhibits the IgE-dependent passive cutaneous anaphylaxis reaction in mice at a concentration of 25 mg/kg; inhibits growth of HRA ovarian and Bxpc-3 pancreatic cancer cells in a dose-dependent manner; decreases immobility time in the forced swim test in a mouse model of colorectal carcinoma; reduces infarct volume in a rat model of ischemia-reperfusion-induced brain injury
Catalog No. 50-298-7855 Supplier Cayman Chemical Supplier No. 2339010 MG
A natural steroid glycoside with diverse effects; inhibits release of β-hexosaminidase from RBL-2H3 cells (IC50 = 100 μM) and inhibits the IgE-dependent passive cutaneous anaphylaxis reaction in mice at a concentration of 25 mg/kg; inhibits growth of HRA ovarian and Bxpc-3 pancreatic cancer cells in a dose-dependent manner; decreases immobility time in the forced swim test in a mouse model of colorectal carcinoma; reduces infarct volume in a rat model of ischemia-reperfusion-induced brain injury