Learn More
Medchemexpress LLC 20(S)-hydroxycholesterol | 516-72-3 | MFCD16661190 | 98.7% | 402.7 g/mol | C27H46O2 | 5MG
SDP
Catalog No. 5000238261
Encompass_Preferred
20(S)-Hydroxycholesterol is an oxysterol used as a research reagent and analytical standard. It acts as an allosteric activator of the Smoothened (Smo) protein in the Hedgehog signaling pathway and is commonly used in developmental biology, oncology, and cell signaling studies. The compound has CAS RN 516-72-3, molecular formula C27H46O2, and molecular weight 402.7 g/mol.
Catalog No. 50-002-38261 Supplier Medchemexpress LLC Supplier No. HY12316R5MG
20(S)-Hydroxycholesterol is an oxysterol used as a research reagent and analytical standard. It acts as an allosteric activator of the Smoothened (Smo) protein in the Hedgehog signaling pathway and is commonly used in developmental biology, oncology, and cell signaling studies. The compound has CAS RN 516-72-3, molecular formula C27H46O2, and molecular weight 402.7 g/mol.