Learn More
Cayman Chemical 20(S)-Protopanaxadiol, Purity: ≥95%, CAS: 30636-90-9, 100 mg
SDP
Catalog No. 502946602
Encompass_Preferred
A sapogenin; cytotoxic to MT-4 human T cell leukemia cells (CC50 = 7.2 µM); inhibits the growth of human A549 human lung carcinoma, SKOV3 ovarian adenocarcinoma, SK-MEL-2 melanoma, and K562 chronic leukemia cells (EC50s = 9.8-20 µM), as well as P388 and L1210 murine leukemia cells (EC50s = 9.8 and 10, respectively); inhibits the growth of H. pylori in vitro (MIC = 50 µg/ml)
Catalog No. 50-294-6602 Supplier Cayman Chemical Supplier No. 33201100 MG
A sapogenin; cytotoxic to MT-4 human T cell leukemia cells (CC50 = 7.2 µM); inhibits the growth of human A549 human lung carcinoma, SKOV3 ovarian adenocarcinoma, SK-MEL-2 melanoma, and K562 chronic leukemia cells (EC50s = 9.8-20 µM), as well as P388 and L1210 murine leukemia cells (EC50s = 9.8 and 10, respectively); inhibits the growth of H. pylori in vitro (MIC = 50 µg/ml)