Learn More
Kd Medical Inc 20% SDS 500ML BOTTLE
SDP
Catalog No. 501018896
Encompass_Preferred
Sodium Dodecyl Sulfate Solution (20%) is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations. One 500ml bottle.
Catalog No. 50-101-8896 Supplier Kd Medical Inc Supplier No. RGE3235
Add to Cart
Add to Cart
Sodium Dodecyl Sulfate Solution (20%) is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations. One 500ml bottle.