Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc SDS (20%), 100 mL
SDP

Catalog No. 507517354
Encompass_Preferred

Sodium Dodecyl Sulfate (SDS) is an anionic detergent with applications in SDS-PAGE protein analysis and nucleic acid purification. It is a wetting agent that is effective in both acid and alkaline solutions, and is most often used as a protein and lipid solubilization reagent. SDS is also a powerful protein denaturant.

Recommendation: If 20% SDS gels at low temperature, warm at 30-40°C until the solution clears. This precipitation will not affect the performance of the product.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7354 Supplier Quality Biological Inc Supplier No. 351066721EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Dodecyl Sulfate (SDS) is an anionic detergent with applications in SDS-PAGE protein analysis and nucleic acid purification. It is a wetting agent that is effective in both acid and alkaline solutions, and is most often used as a protein and lipid solubilization reagent. SDS is also a powerful protein denaturant.

Recommendation: If 20% SDS gels at low temperature, warm at 30-40°C until the solution clears. This precipitation will not affect the performance of the product.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.