Learn More
Teknova 20% SDS Solution. 1000mL, Sterile.
SDP
Catalog No. 50843080
Encompass
Sodium Dodecyl Sulfate (SDS) is an anionic surfactant solution and is one of the most commonly used detergents in protein purification and electrophoretic separation of proteins and lipids. It is often used as an adjuvant in lysing cells during DNA extraction and as a protein denaturing agent in SDS-PAGE.
This item is not returnable. View return policy
Catalog No. 50-843-080 Supplier Teknova Supplier No. S0295
Sodium Dodecyl Sulfate (SDS) is an anionic surfactant solution and is one of the most commonly used detergents in protein purification and electrophoretic separation of proteins and lipids. It is often used as an adjuvant in lysing cells during DNA extraction and as a protein denaturing agent in SDS-PAGE.