Learn More
Teknova 20% SDS Solution. 500 mL, Sterile. 2 pack.
SDP
Catalog No. 502387709
Encompass
Sodium Dodecyl Sulfate (SDS) is an anionic surfactant solution and is one of the most commonly used detergents in protein purification and electrophoretic separation of proteins and lipids. It is often used as an adjuvant in lysing cells during DNA extraction and as a protein denaturing agent in SDS-PAGE.
Catalog No. 50-238-7709 Supplier Teknova Supplier No. S02942PK
Sodium Dodecyl Sulfate (SDS) is an anionic surfactant solution and is one of the most commonly used detergents in protein purification and electrophoretic separation of proteins and lipids. It is often used as an adjuvant in lysing cells during DNA extraction and as a protein denaturing agent in SDS-PAGE.