Learn More
Electron Microscopy Sciences 30 Slide Pop-Up Slide Folder
SDP
Catalog No. 501899627
Encompass_Preferred
Holds 1x3" slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed.
Catalog No. 50-189-9627 Supplier Electron Microscopy Sciences Supplier No. 7152210
Holds 1x3" slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed.