Learn More
Electron Microscopy Sciences Winlab Rack Carrier Blue for 20 Slide Rack
SDP
Catalog No. NC1332214
Encompass_Preferred
Steel wire hooks molded to the polypropylene handle to form slide rack hanger, easy to attach or detach from the WINLAB Slide Racks. Available in: Red, White, Blue, Green and Yellow. Fits all LS-20 and LSM-20 Slide Racks.
Capacity: 20 slides
Catalog No. NC1332214 Supplier Electron Microscopy Sciences Supplier No. 71435B
Steel wire hooks molded to the polypropylene handle to form slide rack hanger, easy to attach or detach from the WINLAB Slide Racks. Available in: Red, White, Blue, Green and Yellow. Fits all LS-20 and LSM-20 Slide Racks.
Capacity: 20 slides