Learn More
Electron Microscopy Sciences StainTray™ 20 Slide Staining Tray With Clear Lid
SDP
Catalog No. 501899564
Encompass_Preferred
An ideal device for immunohistochemistry slide staining. The StainTray has a black base, made from tough ABS plastic, withstanding a wide range of chemicals (except chlorinated hydrocarbons). Features four plastic rails covered with a polymer strip to perfectly hold the slides even if the tray is held at an angle.
When humidity is needed, the wells between the rails will hold the water securely without splashing. Rails are raised not only to avoid water touching the slides but also make the slides more easily retrievable. The base will also hold excess stain solution that drips from the slides. Four rubber feet add to the base for stability. Units are stackable to save space.
This tray is not only suitable for routine staining requiring a humid chamber but is also ideal for hematology, cytology, and microscopy laboratories.
Capacity: 20 slides
Catalog No. 50-189-9564 Supplier Electron Microscopy Sciences Supplier No. 71397C
An ideal device for immunohistochemistry slide staining. The StainTray has a black base, made from tough ABS plastic, withstanding a wide range of chemicals (except chlorinated hydrocarbons). Features four plastic rails covered with a polymer strip to perfectly hold the slides even if the tray is held at an angle.
When humidity is needed, the wells between the rails will hold the water securely without splashing. Rails are raised not only to avoid water touching the slides but also make the slides more easily retrievable. The base will also hold excess stain solution that drips from the slides. Four rubber feet add to the base for stability. Units are stackable to save space.
This tray is not only suitable for routine staining requiring a humid chamber but is also ideal for hematology, cytology, and microscopy laboratories.
Capacity: 20 slides