Learn More
ATCC 20 strain staggered mix; genomic material
Supplier:  ATCC MSA1003
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises genomic DNA prepared from fully sequenced, characterized, and authenticated ATCC Genuine Cultures that were selected based on relevant phenotypic and genotypic attributes, such as Gram stain, GC content, genome size, and spore formation. Each order for this product is provided with access to run 20 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 6.08 ng/μL to 10.14 ng/μL* *DNA concentrations indicate ATCC manufacturing specifications and are provided as a reference only
Catalog No. 50-238-2811
 Restricted Item