Learn More
Thermco Products Inc Precision lab Hg thermometer, -20 deg./110 deg.C, 1.0 deg.C div, 76mm imm, 305mm
Catalog No. NC0676807
Encompass_Preferred
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing. SAMA No. CP15
Catalog No. NC0676807 Supplier Thermco Products Inc Supplier No. ACC6113
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing. SAMA No. CP15