Learn More
Teknova 20% (v/v) Glycerol Solution. 1000 mL. Sterile. 2 pack.
SDP
Catalog No. 502387420
Encompass
Glycerol solution is a versatile solvent that can be used in a variety of applications including electrophoresis, bacterial stock cultures, as an additive as a carbon source, and stabilizer.
Catalog No. 50-238-7420 Supplier Teknova Supplier No. G17232PK
Glycerol solution is a versatile solvent that can be used in a variety of applications including electrophoresis, bacterial stock cultures, as an additive as a carbon source, and stabilizer.