Learn More
Sprayer Depot 20" X 1" WATER SENSITIVE PAPER
SDP
Catalog No. NC0885838
Encompass_Preferred
These specially coated papers are used for evaluating spray distributions, swath widths, droplet densities and penetration of spray. Water sensitive paper is yellow and is stained blue by exposure to aqueous spray droplets.
Catalog No. NC0885838 Supplier Sprayer Depot Supplier No. 203013N
These specially coated papers are used for evaluating spray distributions, swath widths, droplet densities and penetration of spray. Water sensitive paper is yellow and is stained blue by exposure to aqueous spray droplets.