Learn More
MOLDPRO INC Ion Chromatography (IC) Sample Vial w/ pierce cap, ~7 mL, 20 x 49 mm, Clear, 500/case, Flat Bottom, PP Copolymer Vial, LDPE cap
Catalog No. NC1838658
Encompass_Preferred
MoldPro MP-113P IC Sample Vials are a direct replacement for Dionex items 055058 & 074228 and Environmental Express item K4300 for Dionex AS-50 and AS-AP autosamplers.
Catalog No. NC1838658 Supplier MOLDPRO INC Supplier No. MP113P
MoldPro MP-113P IC Sample Vials are a direct replacement for Dionex items 055058 & 074228 and Environmental Express item K4300 for Dionex AS-50 and AS-AP autosamplers.