Learn More
Teknova 20 x MOPS/SDS Running Buffer. 500mL, Sterile. 8 pack.
SDP
Catalog No. 502387762
Encompass
MOPS (3-(N-morpholino)propanesulfonic acid) is a Good’s buffer and has a pKa of 7.01, making it an ideal buffer for many biological systems at near-neutral pH. MOPS can be used in cell culture media and biopharma buffer formulations as well as in the purification of antibodies, peptides, proteins and blood components. It is nontoxic to cell culture lines.
Catalog No. 50-238-7762 Supplier Teknova Supplier No. M10898PK
MOPS (3-(N-morpholino)propanesulfonic acid) is a Good’s buffer and has a pKa of 7.01, making it an ideal buffer for many biological systems at near-neutral pH. MOPS can be used in cell culture media and biopharma buffer formulations as well as in the purification of antibodies, peptides, proteins and blood components. It is nontoxic to cell culture lines.