Learn More
Sentry Air Systems Model 200 Activated Carbon Filter - 2.5 lb
SDP
Catalog No. NC0241947
Encompass
Model 200 Activated Carbon Filter features 2.5 lbs of heavy-duty activated carbon from crushed virgin coconuts. Carbon filters are ideal for removal of gases, fumes, and vapors. Efficiency depends on the application. Fits Model 200 systems.
Catalog No. NC0241947 Supplier Sentry Air Systems Supplier No. SS200CFP
Model 200 Activated Carbon Filter features 2.5 lbs of heavy-duty activated carbon from crushed virgin coconuts. Carbon filters are ideal for removal of gases, fumes, and vapors. Efficiency depends on the application. Fits Model 200 systems.