Learn More
Braintree Scientific 200 Bags only, clear 12.2"" x 26""
SDP
Catalog No. 501954149
Encompass
Benchtop Bag Holder Perfect benchtop disposal container for tips, tubs, gloves, plates, etc. Tailor-made for 8.5" x 11" autoclave bags, providing the perfect disposal container for all the contaminated or expended materials in your lab. Compact 3" x 5" footprint Autoclavable sturdy, steel contruction Biohazard bags feature printed marking area **Sold for research purposes only and not for human use**
Catalog No. 50-195-4149 Supplier Braintree Scientific Supplier No. A9000C
Benchtop Bag Holder Perfect benchtop disposal container for tips, tubs, gloves, plates, etc. Tailor-made for 8.5" x 11" autoclave bags, providing the perfect disposal container for all the contaminated or expended materials in your lab. Compact 3" x 5" footprint Autoclavable sturdy, steel contruction Biohazard bags feature printed marking area **Sold for research purposes only and not for human use**