Learn More
Accuflow Systems Altafluor 200 FEP (Fluoronated Ethylene Propylene) - 200 FEP, 5/16" X 3/8"- 100 FT
Catalog No. 501354996
Encompass_Preferred
Altafluor 200 FEP (Fluoronated Ethylene Propylene) is an economical choice for applications requiring chemical resistance in combination with broad temperature exposure. Altafluor 200 FEP offers excellent clarity which makes it ideal for sight glass/flow monitoring applications. Certified to NSF 51, NSF 61 and NSF 372. It has superior electrical insulation properties and is available in coiled hose and convoluted tubing constructions. Additionally, the outstanding UV transmission rate of FEP makes it a clear choice for UV transmission applications. Altafluor 200 tubing may be used with flare or conventional fittings.
Catalog No. 50-135-4996 Supplier Accuflow Systems Supplier No. 2000375030100
Altafluor 200 FEP (Fluoronated Ethylene Propylene) is an economical choice for applications requiring chemical resistance in combination with broad temperature exposure. Altafluor 200 FEP offers excellent clarity which makes it ideal for sight glass/flow monitoring applications. Certified to NSF 51, NSF 61 and NSF 372. It has superior electrical insulation properties and is available in coiled hose and convoluted tubing constructions. Additionally, the outstanding UV transmission rate of FEP makes it a clear choice for UV transmission applications. Altafluor 200 tubing may be used with flare or conventional fittings.